missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RCBTB1 Full-length ORF (AAH38104.1, 1 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH38104.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55213 |
Molecular Weight (g/mol) | 64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16191283
|
Abnova™
H00055213-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16181283
|
Abnova™
H00055213-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein with an N-terminal RCC1 domain and a C-terminal BTB (broad complex, tramtrack and bric-a-brac) domain. In rat, over-expression of this gene in vascular smooth muscle cells induced cellular hypertrophy. In rat, the C-terminus of RCBTB1 interacts with the angiotensin II receptor-1A. In humans, this gene maps to a region of chromosome 13q that is frequently deleted in B-cell chronic lymphocytic leukemia and other lymphoid malignancies. [provided by RefSeq]
Sequence: MVDVGKWPIFTLLSPQEIASIRKVCVFGTSASEALYVTDNDEVFVFGLNYSNCLGTGDNQSTLVPKKLEGLCGKKIKSLSYGSGPHVLLSTEDGVVYAWGHNGYSQLGNGTTNQGIATVQVCTNLLIKQVVEVACGSHHSMALAADGEVFAWGYNNCGQVGSGSTANQPTPRKVTNCLHIKRVVGIACGQTSSMAVLDNGEVYGWGYNGNGQLGLGNNGNQLTPVRVAALHSVCVNQIVCGYAHTLALTDEGLLYAWGANTYGQLGTGNKNNLLSPAHIMVEKERVVEIAACHSAHTSAAKTQGGHVYMWGQCRGQSVILPHLTHFSCTDDVFACFATPAVSWRLLSVVSIVIGKSpecifications
AAH38104.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64kDa | |
Glutathione Sepharose 4 Fast Flow | |
MVDVGKWPIFTLLSPQEIASIRKVCVFGTSASEALYVTDNDEVFVFGLNYSNCLGTGDNQSTLVPKKLEGLCGKKIKSLSYGSGPHVLLSTEDGVVYAWGHNGYSQLGNGTTNQGIATVQVCTNLLIKQVVEVACGSHHSMALAADGEVFAWGYNNCGQVGSGSTANQPTPRKVTNCLHIKRVVGIACGQTSSMAVLDNGEVYGWGYNGNGQLGLGNNGNQLTPVRVAALHSVCVNQIVCGYAHTLALTDEGLLYAWGANTYGQLGTGNKNNLLSPAHIMVEKERVVEIAACHSAHTSAAKTQGGHVYMWGQCRGQSVILPHLTHFSCTDDVFACFATPAVSWRLLSVVSIVIGK | |
RUO | |
RCBTB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55213 | |
RCBTB1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLLD7/CLLL7/GLP/MGC33184/RP11-185C18.1 | |
RCBTB1 | |
Recombinant | |
wheat germ expression system |