missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RARRES1 Partial ORF (NP_996846, 205 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_996846 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5918 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184125
|
Abnova™
H00005918-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16194125
|
Abnova™
H00005918-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: EMTTQVSHYYLAQLTSVRQWKTNDDTIDFDYTVLLHELSTQEIIPCRIHLVWYPGKPLKVKYHCQELQTPEEASGTEEGSAVVPTELSNFSpecifications
NP_996846 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TIG1 | |
RARRES1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5918 | |
RARRES1 (Human) Recombinant Protein (Q01) | |
EMTTQVSHYYLAQLTSVRQWKTNDDTIDFDYTVLLHELSTQEIIPCRIHLVWYPGKPLKVKYHCQELQTPEEASGTEEGSAVVPTELSNF | |
RUO | |
RARRES1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |