missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RARRES1 Partial ORF (NP_996846, 205 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005918-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16184125

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: EMTTQVSHYYLAQLTSVRQWKTNDDTIDFDYTVLLHELSTQEIIPCRIHLVWYPGKPLKVKYHCQELQTPEEASGTEEGSAVVPTELSNF
Specifications

Specifications

NP_996846
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EMTTQVSHYYLAQLTSVRQWKTNDDTIDFDYTVLLHELSTQEIIPCRIHLVWYPGKPLKVKYHCQELQTPEEASGTEEGSAVVPTELSNF
RUO
RARRES1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5918
RARRES1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TIG1
RARRES1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers