missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PYY ELISA Kit

Human PYY quantitates human PYY in serum, plasma, supernatant.

Brand:  Invitrogen™ EH387RB

Additional Details : Weight : 0.33820kg

 View more versions of this product

Product Code. 17104693

  • 5430.00 NOK / Pack of 96

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

Human PYY quantitates human PYY in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PYY.

Peptide tyrosine-tyrosine amide (3-36), or PYY (3-36), is a major metabolite of the gut hormone PYY and is produced by the action of dipeptidyl peptidase IV on PYY in the intestinal brush border and in the circulation. PYY (3-36) is a potent inhibitor of food intake in rats and humans, acting selectively on the Y2 receptor to inhibit orexigenic NPY neurons in the arcuate nucleus, thus disinhibiting anorexigenic POMC neurons. The anorexigenic effect of PYY (3-36) is additive to that of the co-secreted gut hormone GLP-1.
Specifications

Specifications

GHYY; Peptide tyrosine tyrosine; peptide tyrosine-tyrosine (YY); peptide YY; peptide YY (mapped); Peptide YY(3-36); Peptide YY3-36; Peptide YY3-37; peptide-YY; prepro-PYY; Pyy; PYY II; PYY1; PYY-1; PYY-2; PYYI; PYY-I; PYYII; PYY-II; RATGHYY; Yy
0.15 ng/mL
ELISA
Colorimetric Microplate Reader
Pyy
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
0.15 to 10 ng/mL
Biotin
Serum, Plasma, Supernatant
P10082
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Supernatant, 50 µL; Serum, 50 µL; Plasma, 50 µL
PYY
4 hr 45 min
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers