missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PURG Full-length ORF (NP_037489.1, 1 a.a. - 347 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00029942-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189212

  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The exact function of this gene is not known, however, its encoded product is highly similar to purine-rich element binding protein A. The latter is a DNA-binding protein which binds preferentially to the single strand of the purine-rich element termed PUR, and has been implicated in the control of both DNA replication and transcription. This gene lies in close proximity to the Werner syndrome gene, but on the opposite strand, on chromosome 8p11. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MERARRRGGGGGRGRGGKNVGGSGLSKSRLYPQAQHSHYPHYAASATPNQAGGAAEIQELASKRVDIQKKRFYLDVKQSSRGRFLKIAEVWIGRGRQDNIRKSKLTLSLSVAAELKDCLGDFIEHYAHLGLKGHRQEHGHSKEQGSRRRQKHSAPSPPVSVGSEEHPHSVLKTDYIERDNRKYYLDLKENQRGRFLRIRQTMMRGTGMIGYFGHSLGQEQTIVLPAQGMIEFRDALVQLIEDYGEGDIEERRGGDDDPLELPEGTSFRVDNKRFYFDVGSNKYGIFLKVSEVRPPYRNTITVPFKAWTRFGENFIKYEEEMRKICNSHKEKRMDGRKASGEEQECLD
Specifications

Specifications

NP_037489.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC119274/PURG-A/PURG-B
PURG
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
29942
PURG (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MERARRRGGGGGRGRGGKNVGGSGLSKSRLYPQAQHSHYPHYAASATPNQAGGAAEIQELASKRVDIQKKRFYLDVKQSSRGRFLKIAEVWIGRGRQDNIRKSKLTLSLSVAAELKDCLGDFIEHYAHLGLKGHRQEHGHSKEQGSRRRQKHSAPSPPVSVGSEEHPHSVLKTDYIERDNRKYYLDLKENQRGRFLRIRQTMMRGTGMIGYFGHSLGQEQTIVLPAQGMIEFRDALVQLIEDYGEGDIEERRGGDDDPLELPEGTSFRVDNKRFYFDVGSNKYGIFLKVSEVRPPYRNTITVPFKAWTRFGENFIKYEEEMRKICNSHKEKRMDGRKASGEEQECLD
RUO
PURG
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers