Learn More
Abnova™ Human PURG Full-length ORF (NP_037489.1, 1 a.a. - 347 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_037489.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 29942 |
Molecular Weight (g/mol) | 66kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16199212
|
Abnova™
H00029942-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16189212
|
Abnova™
H00029942-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The exact function of this gene is not known, however, its encoded product is highly similar to purine-rich element binding protein A. The latter is a DNA-binding protein which binds preferentially to the single strand of the purine-rich element termed PUR, and has been implicated in the control of both DNA replication and transcription. This gene lies in close proximity to the Werner syndrome gene, but on the opposite strand, on chromosome 8p11. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MERARRRGGGGGRGRGGKNVGGSGLSKSRLYPQAQHSHYPHYAASATPNQAGGAAEIQELASKRVDIQKKRFYLDVKQSSRGRFLKIAEVWIGRGRQDNIRKSKLTLSLSVAAELKDCLGDFIEHYAHLGLKGHRQEHGHSKEQGSRRRQKHSAPSPPVSVGSEEHPHSVLKTDYIERDNRKYYLDLKENQRGRFLRIRQTMMRGTGMIGYFGHSLGQEQTIVLPAQGMIEFRDALVQLIEDYGEGDIEERRGGDDDPLELPEGTSFRVDNKRFYFDVGSNKYGIFLKVSEVRPPYRNTITVPFKAWTRFGENFIKYEEEMRKICNSHKEKRMDGRKASGEEQECLDSpecifications
NP_037489.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66kDa | |
Glutathione Sepharose 4 Fast Flow | |
MERARRRGGGGGRGRGGKNVGGSGLSKSRLYPQAQHSHYPHYAASATPNQAGGAAEIQELASKRVDIQKKRFYLDVKQSSRGRFLKIAEVWIGRGRQDNIRKSKLTLSLSVAAELKDCLGDFIEHYAHLGLKGHRQEHGHSKEQGSRRRQKHSAPSPPVSVGSEEHPHSVLKTDYIERDNRKYYLDLKENQRGRFLRIRQTMMRGTGMIGYFGHSLGQEQTIVLPAQGMIEFRDALVQLIEDYGEGDIEERRGGDDDPLELPEGTSFRVDNKRFYFDVGSNKYGIFLKVSEVRPPYRNTITVPFKAWTRFGENFIKYEEEMRKICNSHKEKRMDGRKASGEEQECLD | |
RUO | |
PURG | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
29942 | |
PURG (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119274/PURG-A/PURG-B | |
PURG | |
Recombinant | |
wheat germ expression system |