Learn More
R&D Systems™ Human Proprotein Convertase 4 (NP_060043.2) VersaClone cDNA
Product Code. 18724497
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18724497 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18724497 Supplier R&D Systems™ Supplier No. RDC3601
  • ONLINE EXCLUSIVE PRICE
    8439.37 NOK
    7465.50 NOK / 10µg
    Save 973.87 NOK
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Specifications
Gene Proprotein convertase PC4
Accession Number NP_060043.2
Gene ID (Entrez) 54760
Gene Alias DKFZp434B217, EC 3.4.21, EC 3.4.21.75, EC 3.4.21.93, MGC34749, PC4, PC4EC 3.4.21.-, Proprotein convertase 4, proprotein convertase subtilisin/kexin type 4, SPC5
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg