missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PILR-alpha Antibody, R&D Systems™

Rabbit Monoclonal Antibody
1930.00 NOK - 5120.00 NOK
Specifications
| Antigen | PILR-alpha |
|---|---|
| Clone | 2175B |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Neutralization 1-5 ug/mL |
| Applications | Flow Cytometry, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18764839
![]() |
R&D Systems
MAB64842-SP |
25 μg |
1930.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700959
![]() |
R&D Systems
MAB64842-100 |
100 μg |
5120.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PILR-alpha Monoclonal specifically detects PILR-alpha in Human samples. It is validated for Flow Cytometry, Neutralization.Specifications
| PILR-alpha | |
| Flow Cytometry 0.25 ug/10^6 cells, Neutralization 1-5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha | |
| Mouse myeloma cell line NS0-derived recombinant human PILR-alpha with a C-terminal 6-His tag Gln20-Thr196 Accession # Q9UKJ1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PILR-alpha in direct ELISAs. |
| 2175B | |
| Flow Cytometry, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UKJ1 | |
| 29992 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title