missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PILR-alpha Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB64842-SP
This item is not returnable.
View return policy
Description
PILR-alpha Monoclonal specifically detects PILR-alpha in Human samples. It is validated for Flow Cytometry, Neutralization.
Specifications
| PILR-alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha | |
| Mouse myeloma cell line NS0-derived recombinant human PILR-alpha with a C-terminal 6-His tag Gln20-Thr196 Accession # Q9UKJ1 | |
| 25 μg | |
| Primary | |
| Detects human PILR-alpha in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Neutralization | |
| 2175B | |
| Flow Cytometry 0.25 ug/10^6 cells, Neutralization 1-5 ug/mL | |
| Q9UKJ1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 29992 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction