missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PGBD2 Full-length ORF (NP_733843.1, 1 a.a. - 592 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00267002-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189793

  • 4115.00 NOK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. The exact function of this gene is not known. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MASTSRDVIAGRGIHSKVKSAKLLEVLNAMEEEESNNNREEIFIAPPDNAAGEFTDEDSGDEDSQRGAHLPGSVLHASVLCEDSGTGEDNDDLELQPAKKRQKAVVKPQRIWTKRDIRPDFGSWTASDPHIEDLKSQELSPVGLFELFFDEGTINFIVNETNRYAWQKNVNLSLTAQELKCVLGILILSGYISYPRRRMFWETSPDSHHHLVADAIRRDRFELIFSYLHFADNNELDASDRFAKVRPLIIRMNCNFQKHAPLEEFYSFGESMCEYFGHRGSKQLHRGKPVRLGYKIWCGTTSRGYLVWFEPSQGTLFTKPDRSLDLGGSMVIKFVDALQERGFLPYHIFFDKVFTSVKLMSILRKKGVKATGTVREYRTERCPLKDPKELKKMKRGSFDYKVDESEEIIVCRWHDSSVVNICSNAVGIEPVRLTSRHSGAAKTRTQVHQPSLVKLYQEKVGGVGRMDQNIAKYKVKIRGMKWYSSFIGYVIDAALNNAWQLHRICCQDAQVDLLAFRRYIACVYLESNADTTSQGRRSRRLETESRFDMIGHWIIHQDKRTRCALCHSQTNTRCEKCQKGVHAKCFREYHIR
Specifications

Specifications

NP_733843.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
94.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PGBD2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
267002
PGBD2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASTSRDVIAGRGIHSKVKSAKLLEVLNAMEEEESNNNREEIFIAPPDNAAGEFTDEDSGDEDSQRGAHLPGSVLHASVLCEDSGTGEDNDDLELQPAKKRQKAVVKPQRIWTKRDIRPDFGSWTASDPHIEDLKSQELSPVGLFELFFDEGTINFIVNETNRYAWQKNVNLSLTAQELKCVLGILILSGYISYPRRRMFWETSPDSHHHLVADAIRRDRFELIFSYLHFADNNELDASDRFAKVRPLIIRMNCNFQKHAPLEEFYSFGESMCEYFGHRGSKQLHRGKPVRLGYKIWCGTTSRGYLVWFEPSQGTLFTKPDRSLDLGGSMVIKFVDALQERGFLPYHIFFDKVFTSVKLMSILRKKGVKATGTVREYRTERCPLKDPKELKKMKRGSFDYKVDESEEIIVCRWHDSSVVNICSNAVGIEPVRLTSRHSGAAKTRTQVHQPSLVKLYQEKVGGVGRMDQNIAKYKVKIRGMKWYSSFIGYVIDAALNNAWQLHRICCQDAQVDLLAFRRYIACVYLESNADTTSQGRRSRRLETESRFDMIGHWIIHQDKRTRCALCHSQTNTRCEKCQKGVHAKCFREYHIR
RUO
PGBD2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers