Learn More
Abnova™ Human PGBD2 Full-length ORF (NP_733843.1, 1 a.a. - 592 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_733843.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 267002 |
Molecular Weight (g/mol) | 94.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16189793
|
Abnova™
H00267002-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16199793
|
Abnova™
H00267002-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. The exact function of this gene is not known. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MASTSRDVIAGRGIHSKVKSAKLLEVLNAMEEEESNNNREEIFIAPPDNAAGEFTDEDSGDEDSQRGAHLPGSVLHASVLCEDSGTGEDNDDLELQPAKKRQKAVVKPQRIWTKRDIRPDFGSWTASDPHIEDLKSQELSPVGLFELFFDEGTINFIVNETNRYAWQKNVNLSLTAQELKCVLGILILSGYISYPRRRMFWETSPDSHHHLVADAIRRDRFELIFSYLHFADNNELDASDRFAKVRPLIIRMNCNFQKHAPLEEFYSFGESMCEYFGHRGSKQLHRGKPVRLGYKIWCGTTSRGYLVWFEPSQGTLFTKPDRSLDLGGSMVIKFVDALQERGFLPYHIFFDKVFTSVKLMSILRKKGVKATGTVREYRTERCPLKDPKELKKMKRGSFDYKVDESEEIIVCRWHDSSVVNICSNAVGIEPVRLTSRHSGAAKTRTQVHQPSLVKLYQEKVGGVGRMDQNIAKYKVKIRGMKWYSSFIGYVIDAALNNAWQLHRICCQDAQVDLLAFRRYIACVYLESNADTTSQGRRSRRLETESRFDMIGHWIIHQDKRTRCALCHSQTNTRCEKCQKGVHAKCFREYHIRSpecifications
NP_733843.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
94.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MASTSRDVIAGRGIHSKVKSAKLLEVLNAMEEEESNNNREEIFIAPPDNAAGEFTDEDSGDEDSQRGAHLPGSVLHASVLCEDSGTGEDNDDLELQPAKKRQKAVVKPQRIWTKRDIRPDFGSWTASDPHIEDLKSQELSPVGLFELFFDEGTINFIVNETNRYAWQKNVNLSLTAQELKCVLGILILSGYISYPRRRMFWETSPDSHHHLVADAIRRDRFELIFSYLHFADNNELDASDRFAKVRPLIIRMNCNFQKHAPLEEFYSFGESMCEYFGHRGSKQLHRGKPVRLGYKIWCGTTSRGYLVWFEPSQGTLFTKPDRSLDLGGSMVIKFVDALQERGFLPYHIFFDKVFTSVKLMSILRKKGVKATGTVREYRTERCPLKDPKELKKMKRGSFDYKVDESEEIIVCRWHDSSVVNICSNAVGIEPVRLTSRHSGAAKTRTQVHQPSLVKLYQEKVGGVGRMDQNIAKYKVKIRGMKWYSSFIGYVIDAALNNAWQLHRICCQDAQVDLLAFRRYIACVYLESNADTTSQGRRSRRLETESRFDMIGHWIIHQDKRTRCALCHSQTNTRCEKCQKGVHAKCFREYHIR | |
RUO | |
PGBD2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
267002 | |
PGBD2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PGBD2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |