Learn More
R&D Systems™ Human PELI1/Pellino-1 (NP_065702.2) VersaClone cDNA
Specifications
Specifications
| Gene | Pellino 1 |
| Gene ID (Entrez) | 57162 |
| Gene Alias | DKFZp686C18116, MGC50990, pellino (Drosophila) homolog 1, pellino homolog 1 (Drosophila), pellino-1, Pellino-related intracellular-signaling molecule, PRISM, protein pellino homolog 1 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 10 μg |