Learn More
R&D Systems™ Human PDGF-AA XL Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
100 tests
Specifications
Specifications
| Product Type | Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform |
| Quantity | 1 Kit |
| Species | Human |
| Binding Target | PDGF-AB/BB |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |