Learn More
Abnova™ Human NOXA1 Full-length ORF (NP_006638.1, 1 a.a. - 483 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010811-P01.10ug
Additional Details : Weight : 0.00010kg
Description
NADPH oxidases (NOXs) catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species (ROS). The NOX activator NOXA1 can stimulate both NOX1 (MIM 300225) and NOX2 (CYBB; MIM 300481), but it appears to be more effective in activating NOX1 (Opitz et al., 2007 [PubMed 17189823]).[supplied by OMIM]
Sequence: MASLGDLVRAWHLGAQAVDRGDWARALHLFSGVPAPPARLCFNAGCVHLLAGDPEAALRAFDQAVTKDTCMAVGFFQRGVANFQLARFQEALSDFWLALEQLRGHAAIDYTQLGLRFKLQAWEVLHNVASAQCQLGLWTEAASSLREAMSKWPEGSLNGLDSALDQVQRRGSLPPRQVPRGEVFRPHRWHLKHLEPVDFLGKAKVVASAIPDDQGWGVRPQQPQGPGANHDARSLIMDSPRAGTHQGPLDAETEVGADRCTSTAYQEQRPQVEQVGKQAPLSPGLPAMGGPGPGPCEDPAGAGGAGAGGSEPLVTVTVQCAFTVALRARRGADLSSLRALLGQALPHQAQLGQLSYLAPGEDGHWVPIPEEESLQRAWQDAAACPRGLQLQCRGAGGRPVLYQVVAQHSYSAQGPEDLGFRQGDTVDVLCEEPDVPLAVDQAWLEGHCDGRIGIFPKCFVVPAGPRMSGAPGRLPRSQQGDQPSpecifications
NP_006638.1 | |
Liquid | |
10811 | |
NOXA1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASLGDLVRAWHLGAQAVDRGDWARALHLFSGVPAPPARLCFNAGCVHLLAGDPEAALRAFDQAVTKDTCMAVGFFQRGVANFQLARFQEALSDFWLALEQLRGHAAIDYTQLGLRFKLQAWEVLHNVASAQCQLGLWTEAASSLREAMSKWPEGSLNGLDSALDQVQRRGSLPPRQVPRGEVFRPHRWHLKHLEPVDFLGKAKVVASAIPDDQGWGVRPQQPQGPGANHDARSLIMDSPRAGTHQGPLDAETEVGADRCTSTAYQEQRPQVEQVGKQAPLSPGLPAMGGPGPGPCEDPAGAGGAGAGGSEPLVTVTVQCAFTVALRARRGADLSSLRALLGQALPHQAQLGQLSYLAPGEDGHWVPIPEEESLQRAWQDAAACPRGLQLQCRGAGGRPVLYQVVAQHSYSAQGPEDLGFRQGDTVDVLCEEPDVPLAVDQAWLEGHCDGRIGIFPKCFVVPAGPRMSGAPGRLPRSQQGDQP | |
RUO | |
NOXA1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ25475/MGC131800/NY-CO-31/SDCCAG31/p51NOX | |
NOXA1 | |
Yes | |
wheat germ expression system |