missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NOXA1 Full-length ORF (NP_006638.1, 1 a.a. - 483 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_006638.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10811 |
Molecular Weight (g/mol) | 78.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16105742
|
Abnova™
H00010811-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16195732
|
Abnova™
H00010811-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NADPH oxidases (NOXs) catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species (ROS). The NOX activator NOXA1 can stimulate both NOX1 (MIM 300225) and NOX2 (CYBB; MIM 300481), but it appears to be more effective in activating NOX1 (Opitz et al., 2007 [PubMed 17189823]).[supplied by OMIM]
Sequence: MASLGDLVRAWHLGAQAVDRGDWARALHLFSGVPAPPARLCFNAGCVHLLAGDPEAALRAFDQAVTKDTCMAVGFFQRGVANFQLARFQEALSDFWLALEQLRGHAAIDYTQLGLRFKLQAWEVLHNVASAQCQLGLWTEAASSLREAMSKWPEGSLNGLDSALDQVQRRGSLPPRQVPRGEVFRPHRWHLKHLEPVDFLGKAKVVASAIPDDQGWGVRPQQPQGPGANHDARSLIMDSPRAGTHQGPLDAETEVGADRCTSTAYQEQRPQVEQVGKQAPLSPGLPAMGGPGPGPCEDPAGAGGAGAGGSEPLVTVTVQCAFTVALRARRGADLSSLRALLGQALPHQAQLGQLSYLAPGEDGHWVPIPEEESLQRAWQDAAACPRGLQLQCRGAGGRPVLYQVVAQHSYSAQGPEDLGFRQGDTVDVLCEEPDVPLAVDQAWLEGHCDGRIGIFPKCFVVPAGPRMSGAPGRLPRSQQGDQPSpecifications
NP_006638.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MASLGDLVRAWHLGAQAVDRGDWARALHLFSGVPAPPARLCFNAGCVHLLAGDPEAALRAFDQAVTKDTCMAVGFFQRGVANFQLARFQEALSDFWLALEQLRGHAAIDYTQLGLRFKLQAWEVLHNVASAQCQLGLWTEAASSLREAMSKWPEGSLNGLDSALDQVQRRGSLPPRQVPRGEVFRPHRWHLKHLEPVDFLGKAKVVASAIPDDQGWGVRPQQPQGPGANHDARSLIMDSPRAGTHQGPLDAETEVGADRCTSTAYQEQRPQVEQVGKQAPLSPGLPAMGGPGPGPCEDPAGAGGAGAGGSEPLVTVTVQCAFTVALRARRGADLSSLRALLGQALPHQAQLGQLSYLAPGEDGHWVPIPEEESLQRAWQDAAACPRGLQLQCRGAGGRPVLYQVVAQHSYSAQGPEDLGFRQGDTVDVLCEEPDVPLAVDQAWLEGHCDGRIGIFPKCFVVPAGPRMSGAPGRLPRSQQGDQP | |
RUO | |
NOXA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10811 | |
NOXA1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ25475/MGC131800/NY-CO-31/SDCCAG31/p51NOX | |
NOXA1 | |
Recombinant | |
wheat germ expression system |