missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NAB1 Full-length ORF (AAH35724, 1 a.a. - 487 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004664-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16128131

  • 5540.00 NOK / 25µg
Estimated Shipment: 16-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MAAALPRTLGELQLYRILQKANLLSYFDAFIQQGGDDVQQLCEAGEEEFLEIMALVGMASKPLHVRRLQKALRDWVTNPGLFNQPLTSLPVSSIPIYKLPEGSPTWLGISCSSYERSSNAREPHLKIPKCAATTCVQSLGQGKSDVVGSLALQSVGESRLWQGHHATESEHSLSPADLGSPASPKESSEALDAAAALSVAECVERMAPTLPKSDLNEVKELLKTNKKLAKMIGHIFEMNDDDPHKEEEIRKYSAIYGRFDSKRKDGKHLTLHELTVNEAAAQLCVKDNALLTRRDELFALARQISREVTYKYTYRTTKSKCGERDELSPKRIKVEDGFPDFQDSVQTLFQQARAKSEELAALSSQQPVKVMAKQMEFLCNQAGYERLQHAERRLSAGLYRQSSEEHSPNGLTSDNSDGQGERPLNLRMPNLQNRQPHHFVVDGELSRLYPSEAKSHSSESLGILKDYPHSAFTLEKKVIKTEPEDSR
Specifications

Specifications

AAH35724
Liquid
4664
Human NAB1 Full-length ORF (AAH35724, 1 a.a. - 487 a.a.) Recombinant Protein with GST-tag at N-terminal
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at −80°C. Aliquot to avoid repeated freezing and thawing.
NAB1
Wheat Germ (in vitro)
GST Tag
AP, Array, ELISA, WB-Re
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
79.31kDa
Glutathione Sepharose 4 Fast Flow
25μg
RUO
NAB1
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers