missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NAB1 Full-length ORF (AAH35724, 1 a.a. - 487 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH35724 |
---|---|
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4664 |
Molecular Weight (g/mol) | 79.31kDa |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128131
|
Abnova™
H00004664-P01.25UG |
25μg |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16118131
|
Abnova™
H00004664-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: MAAALPRTLGELQLYRILQKANLLSYFDAFIQQGGDDVQQLCEAGEEEFLEIMALVGMASKPLHVRRLQKALRDWVTNPGLFNQPLTSLPVSSIPIYKLPEGSPTWLGISCSSYERSSNAREPHLKIPKCAATTCVQSLGQGKSDVVGSLALQSVGESRLWQGHHATESEHSLSPADLGSPASPKESSEALDAAAALSVAECVERMAPTLPKSDLNEVKELLKTNKKLAKMIGHIFEMNDDDPHKEEEIRKYSAIYGRFDSKRKDGKHLTLHELTVNEAAAQLCVKDNALLTRRDELFALARQISREVTYKYTYRTTKSKCGERDELSPKRIKVEDGFPDFQDSVQTLFQQARAKSEELAALSSQQPVKVMAKQMEFLCNQAGYERLQHAERRLSAGLYRQSSEEHSPNGLTSDNSDGQGERPLNLRMPNLQNRQPHHFVVDGELSRLYPSEAKSHSSESLGILKDYPHSAFTLEKKVIKTEPEDSRSpecifications
AAH35724 | |
4664 | |
Glutathione Sepharose 4 Fast Flow | |
RUO | |
NAB1 | |
Recombinant | |
Liquid |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.31kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NAB1 | |
Wheat Germ (in vitro) | |
wheat germ expression system |