Learn More
Abnova™ Human MRPL40 Partial ORF (NP_003767.2, 1 a.a. - 51 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064976-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Deletions in this gene may contribute to the etiology of velo-cardio-facial syndrome and DiGeorge syndrome. [provided by RefSeq]
Sequence: MTASVLRSISLALRPTSGLLGTWQTQLRETHQRASLLSFWELIPMRSEPLRSpecifications
NP_003767.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.35kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTASVLRSISLALRPTSGLLGTWQTQLRETHQRASLLSFWELIPMRSEPLR | |
RUO | |
MRPL40 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64976 | |
MRPL40 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ41774/MGC9400/MRP-L22/NLVCF/URIM | |
MRPL40 | |
Recombinant | |
wheat germ expression system |