Learn More
Abnova™ Human MRPL40 Partial ORF (NP_003767.2, 1 a.a. - 51 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_003767.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 64976 |
Molecular Weight (g/mol) | 31.35kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139776
|
Abnova™
H00064976-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16149776
|
Abnova™
H00064976-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Deletions in this gene may contribute to the etiology of velo-cardio-facial syndrome and DiGeorge syndrome. [provided by RefSeq]
Sequence: MTASVLRSISLALRPTSGLLGTWQTQLRETHQRASLLSFWELIPMRSEPLRSpecifications
NP_003767.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.35kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ41774/MGC9400/MRP-L22/NLVCF/URIM | |
MRPL40 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
64976 | |
MRPL40 (Human) Recombinant Protein (Q01) | |
MTASVLRSISLALRPTSGLLGTWQTQLRETHQRASLLSFWELIPMRSEPLR | |
RUO | |
MRPL40 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |