Learn More
ACROBiosystems Human MERTK / Mer Protein, His Tag
Brand: ACROBiosystems MEK-H52H6-1mg
Additional Details : Weight : 0.00010kg
Description
Tyrosine-protein kinase Mer(MERTK) is a member of the TYRO3/AXL/MER (TAM) receptor kinase family and encodes a transmembrane protein with two fibronectin type-III domains, two Ig-like C2-type (immunoglobulin-like) domains, and one tyrosine kinase domain. Following activation by ligand, interacts with GRB2 or PLCG2 and induces phosphorylation of MAPK1, MAPK2, FAK/PTK2 or RAC1. MERTK signaling plays a role in various processes such as macrophage clearance of apoptotic cells, platelet aggregation, cytoskeleton reorganization and engulfment. Functions in the retinal pigment epithelium (RPE) as a regulator of rod outer segments fragments phagocytosis. Plays also an important role in inhibition of Toll-like receptors (TLRs)-mediated innate immune response by activating STAT1, which selectively induces production of suppressors of cytokine signaling SOCS1 and SOCS3.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.5 kDa. The protein migrates as 70-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_006334.2 | |
1 mg | |
Ala 21 - Ile 505 | |
MERTK,Mer | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
54.5 kDa | |
Human MERTK, His Tag (MEK-H52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Ile 505 (Accession # Q12866-1). | |
-20°C | |
MERTK | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Enzymes |