Learn More
Human MCL1 knockout HEK-293T cell lysate, 1 Kit
Product Code. 32153226 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32153226 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32153226 Supplier Abcam Supplier No. ab2569861Kit
 Restricted Item
Estimated Shipment: 07-12-2026
Human MCL1 knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: MCL1 knockout HEK-293T
Spezifikation
For Use With (Application) Western Blot