Learn More
Abnova™ Human LY9 Partial ORF (NP_002339, 156 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004063-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
LY9 belongs to the SLAM family of immunomodulatory receptors (see SLAMF1; MIM 603492) and interacts with the adaptor molecule SAP (SH2D1A; MIM 300490) (Graham et al., 2006 [PubMed 16365421]).[supplied by OMIM]
Sequence: EPQVTMKSVKVSENFSCNITLMCSVKGAEKSVLYSWTPREPHASESNGGSILTVSRTPCDPDLPYICTAQNPVSQRSSLPVHVGQFCTDPGASRGGTTSpecifications
NP_002339 | |
Liquid | |
4063 | |
LY9 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD229/SLAMF3/hly9/mLY9 | |
LY9 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EPQVTMKSVKVSENFSCNITLMCSVKGAEKSVLYSWTPREPHASESNGGSILTVSRTPCDPDLPYICTAQNPVSQRSSLPVHVGQFCTDPGASRGGTT | |
RUO | |
LY9 | |
Wheat Germ (in vitro) | |
GST |