missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LY9 Partial ORF (NP_002339, 156 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_002339 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4063 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100815
|
Abnova™
H00004063-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190805
|
Abnova™
H00004063-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
LY9 belongs to the SLAM family of immunomodulatory receptors (see SLAMF1; MIM 603492) and interacts with the adaptor molecule SAP (SH2D1A; MIM 300490) (Graham et al., 2006 [PubMed 16365421]).[supplied by OMIM]
Sequence: EPQVTMKSVKVSENFSCNITLMCSVKGAEKSVLYSWTPREPHASESNGGSILTVSRTPCDPDLPYICTAQNPVSQRSSLPVHVGQFCTDPGASRGGTTSpecifications
NP_002339 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD229/SLAMF3/hly9/mLY9 | |
LY9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4063 | |
LY9 (Human) Recombinant Protein (Q01) | |
EPQVTMKSVKVSENFSCNITLMCSVKGAEKSVLYSWTPREPHASESNGGSILTVSRTPCDPDLPYICTAQNPVSQRSSLPVHVGQFCTDPGASRGGTT | |
RUO | |
LY9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |