missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems Human Luminex™ Discovery Assay
Click to view available options
Quantity:
1 Plex
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
41 Plex
42 Plex
43 Plex
44 Plex
45 Plex
46 Plex
47 Plex
48 Plex
49 Plex
5 Plex
50 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
Specifications
Specifications
| Quantity | 24 Plex |
| Species | Human |
| Binding Target | Multiplex Cytokine Assay Kits |
| Gene Symbol | GZMB |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction