missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LRRK1 Partial ORF (AAH05408, 560 a.a. - 659 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH05408 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79705 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184373
|
Abnova™
H00079705-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16194373
|
Abnova™
H00079705-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: PGAASDRSEHDLTPMDGETFSQHLQAVKILAVRDLIWVPRRGGDVIVIGLEKDSEAQRGRVIAVLKARELTPHGIMPVSSVKVCWAGWPVRDMVYMAAVMSpecifications
AAH05408 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23119/FLJ27465/KIAA1790/RIPK6/Roco1 | |
LRRK1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
79705 | |
LRRK1 (Human) Recombinant Protein (Q01) | |
PGAASDRSEHDLTPMDGETFSQHLQAVKILAVRDLIWVPRRGGDVIVIGLEKDSEAQRGRVIAVLKARELTPHGIMPVSSVKVCWAGWPVRDMVYMAAVM | |
RUO | |
LRRK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |