missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LRRK1 Partial ORF (AAH05408, 560 a.a. - 659 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079705-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194373

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: PGAASDRSEHDLTPMDGETFSQHLQAVKILAVRDLIWVPRRGGDVIVIGLEKDSEAQRGRVIAVLKARELTPHGIMPVSSVKVCWAGWPVRDMVYMAAVM
Specifications

Specifications

AAH05408
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PGAASDRSEHDLTPMDGETFSQHLQAVKILAVRDLIWVPRRGGDVIVIGLEKDSEAQRGRVIAVLKARELTPHGIMPVSSVKVCWAGWPVRDMVYMAAVM
RUO
LRRK1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
79705
LRRK1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ23119/FLJ27465/KIAA1790/RIPK6/Roco1
LRRK1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers