missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KRTAP17-1 Full-length ORF (AAI46561.1, 1 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083902-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125543

  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This protein is a member of the keratin-associated protein (KAP) family. The KAP proteins form a matrix of keratin intermediate filaments which contribute to the structure of hair fibers. KAP family members appear to have unique, family-specific amino- and carboxyl-terminal regions and are subdivided into three multi-gene families according to amino acid composition: the high sulfur, the ultrahigh sulfur, and the high tyrosine/glycine KAPs. This protein is a member of the ultrahigh sulfur KAP family and the gene is localized to a cluster of KAPs at 17q12-q21. [provided by RefSeq]

Sequence: MGCCPGDCFTCCTQEQNCCEECCCQPGCCGCCGSCCGCGGSGCGGSGCGGSCCGSSCCGSGCGGCGGCGGCGGGCCGSSCCGSSCCGSGCCGPVCCQPTPICDTK
Specifications

Specifications

AAI46561.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KAP17.1/KRTAP16.1/KRTAP17.1
KRTAP17-1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
83902
KRTAP17-1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGCCPGDCFTCCTQEQNCCEECCCQPGCCGCCGSCCGCGGSGCGGSGCGGSCCGSSCCGSGCGGCGGCGGCGGGCCGSSCCGSSCCGSGCCGPVCCQPTPICDTK
RUO
KRTAP17-1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers