Learn More
Abnova™ Human KRTAP17-1 Full-length ORF (AAI46561.1, 1 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAI46561.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 83902 |
Molecular Weight (g/mol) | 38.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115543
|
Abnova™
H00083902-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16125543
|
Abnova™
H00083902-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This protein is a member of the keratin-associated protein (KAP) family. The KAP proteins form a matrix of keratin intermediate filaments which contribute to the structure of hair fibers. KAP family members appear to have unique, family-specific amino- and carboxyl-terminal regions and are subdivided into three multi-gene families according to amino acid composition: the high sulfur, the ultrahigh sulfur, and the high tyrosine/glycine KAPs. This protein is a member of the ultrahigh sulfur KAP family and the gene is localized to a cluster of KAPs at 17q12-q21. [provided by RefSeq]
Sequence: MGCCPGDCFTCCTQEQNCCEECCCQPGCCGCCGSCCGCGGSGCGGSGCGGSCCGSSCCGSGCGGCGGCGGCGGGCCGSSCCGSSCCGSGCCGPVCCQPTPICDTKSpecifications
AAI46561.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGCCPGDCFTCCTQEQNCCEECCCQPGCCGCCGSCCGCGGSGCGGSGCGGSCCGSSCCGSGCGGCGGCGGCGGGCCGSSCCGSSCCGSGCCGPVCCQPTPICDTK | |
RUO | |
KRTAP17-1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
83902 | |
KRTAP17-1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KAP17.1/KRTAP16.1/KRTAP17.1 | |
KRTAP17-1 | |
Recombinant | |
wheat germ expression system |