missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KRT76 Full-length ORF (NP_056932.1, 1 a.a. - 638 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051350-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189932

  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. The type II keratins are clustered in a region of chromosome 12q13. [provided by RefSeq]

Sequence: MNRQVCKKSFSGRSQGFSGRSAVVSGSSRMSCVARSGGAGGGACGFRSGAGSFGSRSLYNLGSNKSISISVAAGSSRAGGFGGGRSSCGFAGGYGGGFGGSYGGGFGGGRGVGSGFGGAGGFGGAGGFGGPGVFGGPGSFGGPGGFGPGGFPGGIQEVIVNQSLLQPLNVEIDPQIGQVKAQEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWELLQQQTTGSGPSSLEPCFESYISFLCKQLDSLLGERGNLEGELKSMQDLVEDFKKKYEDEINKRTAAENEFVGLKKDVDAAFMNKVELQAKVDSLTDEVSFLRTLYEMELSQMQSHASDTSVVLSMDNNRCLDLGSIIAEVRTQYEEIAQRSKSEAEALYQTKLGELQTTAGRHGDDLRNTKSEIMELNRMIQRLRAEIENVKKQNANLQTAIAEAEQRGEMALKDANAKLQDLQTALQKAKDDLARLLRDYQELMNVKLALDVEIATYRKLLEGEECRMSGECQSAVCISVVSNVTSTSGSSGSSRGVFGGVSGSGSGGYKGGSSSSSSSGYGVSGGSGSGYGGVSSGSTGGRGSSGSYQSSSSGSRLGGAGSISVSHSGMGSSSGSIQTSGGSGYKSGGGGSTSIRFSQTTSSSQHSSTK
Specifications

Specifications

NP_056932.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HUMCYT2A/KRT2B/KRT2P
KRT76
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51350
KRT76 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNRQVCKKSFSGRSQGFSGRSAVVSGSSRMSCVARSGGAGGGACGFRSGAGSFGSRSLYNLGSNKSISISVAAGSSRAGGFGGGRSSCGFAGGYGGGFGGSYGGGFGGGRGVGSGFGGAGGFGGAGGFGGPGVFGGPGSFGGPGGFGPGGFPGGIQEVIVNQSLLQPLNVEIDPQIGQVKAQEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWELLQQQTTGSGPSSLEPCFESYISFLCKQLDSLLGERGNLEGELKSMQDLVEDFKKKYEDEINKRTAAENEFVGLKKDVDAAFMNKVELQAKVDSLTDEVSFLRTLYEMELSQMQSHASDTSVVLSMDNNRCLDLGSIIAEVRTQYEEIAQRSKSEAEALYQTKLGELQTTAGRHGDDLRNTKSEIMELNRMIQRLRAEIENVKKQNANLQTAIAEAEQRGEMALKDANAKLQDLQTALQKAKDDLARLLRDYQELMNVKLALDVEIATYRKLLEGEECRMSGECQSAVCISVVSNVTSTSGSSGSSRGVFGGVSGSGSGGYKGGSSSSSSSGYGVSGGSGSGYGGVSSGSTGGRGSSGSYQSSSSGSRLGGAGSISVSHSGMGSSSGSIQTSGGSGYKSGGGGSTSIRFSQTTSSSQHSSTK
RUO
KRT76
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers