missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KRT76 Full-length ORF (NP_056932.1, 1 a.a. - 638 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_056932.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51350 |
Molecular Weight (g/mol) | 92.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16189932
|
Abnova™
H00051350-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16179932
|
Abnova™
H00051350-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. The type II keratins are clustered in a region of chromosome 12q13. [provided by RefSeq]
Sequence: MNRQVCKKSFSGRSQGFSGRSAVVSGSSRMSCVARSGGAGGGACGFRSGAGSFGSRSLYNLGSNKSISISVAAGSSRAGGFGGGRSSCGFAGGYGGGFGGSYGGGFGGGRGVGSGFGGAGGFGGAGGFGGPGVFGGPGSFGGPGGFGPGGFPGGIQEVIVNQSLLQPLNVEIDPQIGQVKAQEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWELLQQQTTGSGPSSLEPCFESYISFLCKQLDSLLGERGNLEGELKSMQDLVEDFKKKYEDEINKRTAAENEFVGLKKDVDAAFMNKVELQAKVDSLTDEVSFLRTLYEMELSQMQSHASDTSVVLSMDNNRCLDLGSIIAEVRTQYEEIAQRSKSEAEALYQTKLGELQTTAGRHGDDLRNTKSEIMELNRMIQRLRAEIENVKKQNANLQTAIAEAEQRGEMALKDANAKLQDLQTALQKAKDDLARLLRDYQELMNVKLALDVEIATYRKLLEGEECRMSGECQSAVCISVVSNVTSTSGSSGSSRGVFGGVSGSGSGGYKGGSSSSSSSGYGVSGGSGSGYGGVSSGSTGGRGSSGSYQSSSSGSRLGGAGSISVSHSGMGSSSGSIQTSGGSGYKSGGGGSTSIRFSQTTSSSQHSSTKSpecifications
NP_056932.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
92.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNRQVCKKSFSGRSQGFSGRSAVVSGSSRMSCVARSGGAGGGACGFRSGAGSFGSRSLYNLGSNKSISISVAAGSSRAGGFGGGRSSCGFAGGYGGGFGGSYGGGFGGGRGVGSGFGGAGGFGGAGGFGGPGVFGGPGSFGGPGGFGPGGFPGGIQEVIVNQSLLQPLNVEIDPQIGQVKAQEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWELLQQQTTGSGPSSLEPCFESYISFLCKQLDSLLGERGNLEGELKSMQDLVEDFKKKYEDEINKRTAAENEFVGLKKDVDAAFMNKVELQAKVDSLTDEVSFLRTLYEMELSQMQSHASDTSVVLSMDNNRCLDLGSIIAEVRTQYEEIAQRSKSEAEALYQTKLGELQTTAGRHGDDLRNTKSEIMELNRMIQRLRAEIENVKKQNANLQTAIAEAEQRGEMALKDANAKLQDLQTALQKAKDDLARLLRDYQELMNVKLALDVEIATYRKLLEGEECRMSGECQSAVCISVVSNVTSTSGSSGSSRGVFGGVSGSGSGGYKGGSSSSSSSGYGVSGGSGSGYGGVSSGSTGGRGSSGSYQSSSSGSRLGGAGSISVSHSGMGSSSGSIQTSGGSGYKSGGGGSTSIRFSQTTSSSQHSSTK | |
RUO | |
KRT76 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51350 | |
KRT76 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUMCYT2A/KRT2B/KRT2P | |
KRT76 | |
Recombinant | |
wheat germ expression system |