missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human JAM-C ELISA Kit
Description
Human JAM-C quantitates human JAM-C in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human JAM-C.
JAM3 is involved in tight junctions and represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
Specifications
Specifications
| Accession Number | 1110002N23Rik; JAM-2; JAM3; JAM-3; JAMC; JAM-C; Jcam3; junction adhesion molecule 3; junction cell adhesion molecule 3; junction cell adhesion molecule C; junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C; UNQ859/PRO1868 |
| Assay Range | 0.12 to 30 ng/mL |
| Assay Sensitivity | 0.12 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 83700 |
| Gene Symbol | JAM3 |
| Interassay CV | <12% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?