Learn More
Human JAB1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6390-SP
This item is not returnable.
View return policy
Description
JAB1 Monoclonal specifically detects JAB1 in Human samples. It is validated for Western Blot.
Specifications
| JAB1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| COP9 constitutive photomorphogenic homolog subunit 5 (Arabidopsis), COP9 signalosome complex subunit 5, COPS5, CSN5, CSN5COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 5, EC 3.4, JAB138 kDa Mov34 homolog, Jun activation domain-binding protein 1, MOV-34, SGN5, SGN5MGC3149, Signalosome subunit 5 | |
| E. coli-derived recombinant human JAB1 Asn158-Ser334 Accession # Q92905 | |
| 25 μg | |
| Primary | |
| Detects human JAB1 in direct ELISAs and Western blots. | |
| Human | |
| IgG2b |
| Western Blot | |
| 687409 | |
| Western Blot 1 ug/mL | |
| Q92905 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10987 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |