Learn More
Human JAB1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB6390-SP
 View more versions of this product
Product Code. 18739148
  • 1665.00 NOK / 25µg
Description
JAB1 Monoclonal specifically detects JAB1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
JAB1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
COP9 constitutive photomorphogenic homolog subunit 5 (Arabidopsis), COP9 signalosome complex subunit 5, COPS5, CSN5, CSN5COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 5, EC 3.4, JAB138 kDa Mov34 homolog, Jun activation domain-binding protein 1, MOV-34, SGN5, SGN5MGC3149, Signalosome subunit 5
E. coli-derived recombinant human JAB1 Asn158-Ser334 Accession # Q92905
25 μg
Primary
Detects human JAB1 in direct ELISAs and Western blots.
Human
IgG2b
Western Blot
687409
Western Blot 1 ug/mL
Q92905
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
10987
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.