missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-31, AOF Recombinant Protein
E. coli Recombinant Protein
Brand: Invitrogen™ RP87715
Additional Details : Weight : 0.01000kg
Description
Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.- Purity: >95% by SDS-Page
- Endotoxin Concentration: AX41 ≤EUs/μg
- Protein Length: 141 aa.
- Biological Activity: <25 ng/mL
- Molecular Weight: 15.8 kDa
- Animal Origin Free: Yes
Specifications
Q6EBC2 | |
20 mM sodium phosphate with 50 mM NaCl and no preservative | |
15.8 kDa | |
1 mg | |
-20°C | |
1700013B14Rik; Il31; IL-31; ILN; Interleukin; interleukin 31; Interleukin-31 | |
Human | |
Recombinant | |
>95% |
Control, ELISA, Functional Assay, Western Blot | |
386653 | |
Human IL-31, AOF | |
E. coli | |
RUO | |
Il31 | |
Unconjugated | |
Lyophilized |