Learn More
Invitrogen™ IL-23 Human ELISA Kit
Engineered for accurate and precise measurement of IL-23 protein levels from samples including serum, plasma and cell culture supernatants. The use of a p19-specific capture antibody and a p40-specific detection antibody renders this sandwich ELISA exquisitely specific for human IL-23.
5730.00 NOK - 39140.00 NOK
Specifications
Accession Number | Q9NPF7 |
---|---|
Assay Range | 15.6-2,000 pg/mL |
Assay Sensitivity | 4.0 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-23
Biotin-Conjugate anti-human IL-23 monoclonal antibody
Avidin-HRP
Human IL-23 Standard, lyophilized, 4000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15501517
|
Invitrogen™
BMS2023-3TEN |
10 x 96 Tests |
39140.00 NOK
Pack of 10 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
15591507
|
Invitrogen™
BMS2023-3 |
96 Tests |
5730.00 NOK
96 tests |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
The Human Interleukin-23 (Hu IL-23) ELISA quantitates Hu IL-23 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-23. Principle of the method The Human IL-23 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
eBioscience's recombinant IL-23 is produced in baculovirus-infected insect cells as an authentic heterodimer of precursor p19 and p40 subunits using a dual promoter expression system. It is distinct from other available forms of the protein in that it is expressed as a true heterodimer, as opposed to a single-chain, pseudo-heterodimer in which the subunits are joined by an artificial linker.Specifications
Q9NPF7 | |
4.0 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
IL-23,IL-23A,IL23P19,P19,SGRF | |
5.9% | |
HRP | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
JKA3 induced upon T cell activation, SGRF | |
1 hr. 20 min. |
15.6-2,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
51561 | |
6.3% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 45 min. |
For Research Use Only