Learn More
Invitrogen™ Human IL-21 Proquantum Immunoassay Kit
ProQuantum Immunoassay Kit
Brand:  Invitrogen™ A35593
Additional Details : Weight : 0.22720kg
 View more versions of this product
Product Code. 15910483
  • 6790.00 NOK / Each
Estimated Shipment: 02-07-2024
Description
Human IL-21 Proquantum Immunoassay Kit
ProQuantum Immunoassay Kit
Specifications
Specifications
Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5°C to -30°C.
96 Tests