missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgG1 Fc, Recombinant Protein
HEK293 cells Recombinant Protein
670.00 NOK - 2300.00 NOK
Specifications
For Use With (Application) | Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot |
---|---|
Formulation | PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
Name | Human IgG1 Fc |
Source | HEK293 cells |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Name | Formulation | Gene Alias | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Name | Formulation | Gene Alias | Price | Quantity & Availability | ||
17184223
|
Invitrogen™
A42560 |
5 μg | Human IgG1 Fc | PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | IgG; IgG Fc; IgG1; IgG1 heavy chain; Immunoglobulin gamma-1 heavy chain |
670.00 NOK
5µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17194223
|
Invitrogen™
A42561 |
50 μg | Human IgG1 Fc | PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | IgG; IgG Fc; IgG1; IgG1 heavy chain; Immunoglobulin gamma-1 heavy chain |
1140.00 NOK
50µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17174223
|
Invitrogen™
A42559 |
5 x 50 μg | Human IgG1 Fc | PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | IgG; IgG Fc; IgG1; IgG1 heavy chain; Immunoglobulin gamma-1 heavy chain |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
17164223
|
Invitrogen™
A42558 |
5 x 5 μg | Human IgG1 Fc | PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | IgG; IgG Fc; IgG1; IgG1 heavy chain; Immunoglobulin gamma-1 heavy chain |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Immunoglobulin G (IgG) is synthesized and secreted by plasma B cells, and constitutes 75% of serum immunoglobulins in humans. IgG antibodies protect the body against the pathogens by agglutination and immobilization, complement activation, toxin neutralization, as well as the antibody-dependent cell-mediated cytotoxicity (ADCC). Recombinant IgG Fc is suggested to represent a potential anti-inflammatory drug for treatment of human autoimmune diseases.Specifications
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
Human IgG1 Fc | |
-20°C | |
<1 EU/μg | |
Human | |
Recombinant | |
≥ 95% |
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
HEK293 cells | |
RUO | |
IgG; IgG Fc; IgG1; IgG1 heavy chain; Immunoglobulin gamma-1 heavy chain | |
Unconjugated | |
Lyophilized |