missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgG1 Fc, Recombinant Protein
HEK293 cells Recombinant Protein
Brand: Invitrogen™ A42559
Additional Details : Weight : 0.01820kg
Description
Immunoglobulin G (IgG) is synthesized and secreted by plasma B cells, and constitutes 75% of serum immunoglobulins in humans. IgG antibodies protect the body against the pathogens by agglutination and immobilization, complement activation, toxin neutralization, as well as the antibody-dependent cell-mediated cytotoxicity (ADCC). Recombinant IgG Fc is suggested to represent a potential anti-inflammatory drug for treatment of human autoimmune diseases.Specifications
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
Human IgG1 Fc | |
HEK293 cells | |
RUO | |
IgG; IgG Fc; IgG1; IgG1 heavy chain; Immunoglobulin gamma-1 heavy chain | |
Unconjugated | |
Lyophilized |
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
5 x 50 μg | |
-20°C | |
<1 EU/μg | |
Human | |
Recombinant | |
≥ 95% |