missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HM74A/PUMA-G/GPR109A Antibody, R&D Systems™

Rat Monoclonal Antibody
1905.00 NOK - 4700.00 NOK
Specifications
| Antigen | HM74A/PUMA-G/GPR109A/NIACR1 |
|---|---|
| Clone | 245115 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18772717
![]() |
R&D Systems
MAB27601-SP |
25 μg |
1905.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18790967
![]() |
R&D Systems
MAB27601-100 |
100 μg |
4700.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Immunohistochemistry.Specifications
| HM74A/PUMA-G/GPR109A/NIACR1 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| GPR109A, HCA2, HCAR2, hydroxycarboxylic acid receptor 2, Niacr1, Nicotinic Acid Receptor, PUMAG | |
| NS0 mouse myeloma cell line transfected with human HM74A/PUMA-G/GPR109A, Met1-Pro363, Accession # Q8TDS4 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human HM74A/PUMA-G/GPR109A in direct ELISAs. |
| 245115 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| Q8TDS4 | |
| 338442 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title