missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HM74A/PUMA-G/GPR109A Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB27601-SP
This item is not returnable.
View return policy
Description
HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| HM74A/PUMA-G/GPR109A/NIACR1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| GPR109A, HCA2, HCAR2, hydroxycarboxylic acid receptor 2, Niacr1, Nicotinic Acid Receptor, PUMAG | |
| NS0 mouse myeloma cell line transfected with human HM74A/PUMA-G/GPR109A, Met1-Pro363, Accession # Q8TDS4 | |
| 25 μg | |
| Primary | |
| Detects human HM74A/PUMA-G/GPR109A in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 245115 | |
| Immunohistochemistry 5-25 ug/mL | |
| Q8TDS4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 338442 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction