Learn More
R&D Systems™ Human Histone Deacetylase 2 (NP_001518.3) VersaClone cDNA
Specifications
Specifications
| Gene | Histone Deacetylase 2/HDAC2 |
| Accession Number | NP_001518.3 |
| Concentration | 10 μg (at 0.2 μg/μL) |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1480 bp |
| Gene ID (Entrez) | 3066 |
| Gene Alias | EC 3.5.1.98, HD2, HDAC2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |