Learn More
ACROBiosystems Human Glypican 1 / GPC1 Protein, His Tag
Brand: ACROBiosystems GP1-H52H9-1mg
Additional Details : Weight : 0.00010kg
Description
Glypican 1 has been shown to interact with SLIT2. This protein is involved in the misfolding of normal prion proteins in the cell membrane to the infectious prion form. Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 58.0 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_002072.2 | |
1 mg | |
Asp 24 - Ser 530 | |
Glypican 1,GPC1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |
58.0 kDa | |
Human Glypican 1, His Tag (GP1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession # P35052-1). | |
-20°C | |
Glypican 1 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |