missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Glypican 1 / GPC1 Protein, His Tag
5341.70 NOK - 39223.36 NOK
Specifications
Accession Number | NP_002072.2 |
---|---|
Molecular Weight (g/mol) | 58.0 kDa |
Source | Human Glypican 1, His Tag (GP1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession # P35052-1). |
Immunogen | Asp 24 - Ser 530 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16447589
|
ACROBiosystems
GP1-H52H9-100UG |
100 μg |
5341.70 NOK
100µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16457589
|
ACROBiosystems
GP1-H52H9-1MG |
1 mg |
39223.36 NOK
1mg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Glypican 1 has been shown to interact with SLIT2. This protein is involved in the misfolding of normal prion proteins in the cell membrane to the infectious prion form. Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 58.0 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_002072.2 | |
Human Glypican 1, His Tag (GP1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession # P35052-1). | |
-20°C | |
Glypican 1 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
58.0 kDa | |
Asp 24 - Ser 530 | |
Glypican 1,GPC1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |