missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Gas6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
1905.00 NOK - 4575.00 NOK
Specifications
| Antigen | Gas6 |
|---|---|
| Clone | 100130 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730287
![]() |
R&D Systems
MAB8853-SP |
25 μg |
1905.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18753707
|
R&D Systems
MAB8853-100 |
100 μg |
4575.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gas6 Monoclonal specifically detects Gas6 in Human samples. It is validated for ELISA.Specifications
| Gas6 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_000811 | |
| 2621 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 100130 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas6, Asp118-Ala678, Accession # NP_000811 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title