missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Gas6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB8853-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Gas6 Monoclonal specifically detects Gas6 in Human samples. It is validated for ELISA.
Spezifikation
| Gas6 | |
| Monoclonal | |
| Unconjugated | |
| NP_000811 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2621 | |
| Human | |
| Purified |
| ELISA | |
| 100130 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas6, Asp118-Ala678, Accession # NP_000811 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur