Learn More
Invitrogen™ Human FGF-17 ELISA Kit
Human FGF-17 quantitates human FGF-17 in serum, plasma, supernatant.
Brand:  Invitrogen™ EH186RB
Additional Details : Weight : 0.33820kg
 View more versions of this product
Product Code. 17104473
  • 5430.00 NOK / Pack of 96
Estimated Shipment: 30-10-2023
Description
Human FGF-17 quantitates human FGF-17 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FGF-17.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was shown to be prominently expressed in the cerebellum and cortex. The mouse homolog of this gene was localized to specific sites in the midline structures of the forebrain, the midbrain-hindbrain junction, developing skeleton and developing arteries, which suggests a role in central nervous system, bone and vascular development. This gene was referred to as FGF-13 in reference 2, however, its amino acid sequence and chromosomal localization are identical to FGF17.
Specifications
Specifications
FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187
0.25 ng/mL
ELISA
Colorimetric Microplate Reader
FGF17
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
0.25 to 60 ng/mL
Biotin
Serum, Plasma, Supernatant
O60258
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Supernatant, 50 µL; Plasma, 50 µL; Serum, 50 µL
FGF17
4 hr 45 min