Learn More
R&D Systems™ Human FAT10 (NP_006389.2) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | FAT10 |
| Accession Number | NP_006389.2 |
| Gene ID (Entrez) | 10537 |
| Gene Alias | Diubiquitin, FAT10diubiquitin, GABBR1, UBD, UBD-3, ubiquitin D, Ubiquitin-like protein FAT10 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |