Learn More
Abnova™ Human Fam40A Full-Length Orf (Bab55265.1, 1 A.A.-742 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
Brand: Abnova™ H00085369-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MNRKCFEEDFRIHVTDKKWTELDTNQHRTHAMRLLDGLEVTAREKRLKVARAILYVAQGTFGECSSEAEVQSWMRYNIFLLLEVGTFNALVELLNMEIDNSAACSSAVRKPAISLADSTDLRVLLNIMYLIVETVHQECEGDKAEW
Specifications
BAB55265.1 | |
Liquid | |
85369 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14743/KIAA1761/MGC148091/RP4-773N10.1 | |
FAM40A |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
FAM40A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MNRKCFEEDFRIHVTDKKWTELDTNQHRTHAMRLLDGLEVTAREKRLKVARAILYVAQGTFGECSSEAEVQSWMRYNIFLLLEVGTFNALVELLNMEIDNSAACSSAVRKPAISLADSTDLRVLLNIMYLIVETVHQECEGDKAEWRTMRQTFRAELGSPLYNNEPFAIMLFGMVTKFCSGHAPHFPMKKVLLLLWKTVLCTLGGFEELQSMKAEKRSILGLPPLPEDSIKVIRNMRAASPPASASDLIEQQQKRGRREHKALIKQDNLDAFNERDPYKADDSREEEEENDDDNSLEGETFPLERDEVMPPPLQHPQTDRLTCPKGLPWAPKVREKDIEMFLESSRSKFIGYTLGSDTNTVVGLPRPIHESIKTLKQHKYTSIAEVQAQMEEEYLRSPLSGGEEEVEQVPAETLYQGLLPSLPQYMIALLKILLAAAPTSKAKTDSINILADVLPEEMPTTVLQSMKLGVDVNRHKEVIVKAISAVLLLLLKHFKLNHVYQFEYMAQHLVFANCIPLILKFFNQNIMSYITAKNSISVLDYPHCVVHELPELTAESLEAGDSNQFCWRNLFSCINLLRILNKLTKWKHSRTMMLVVFKSAPILKRALKVKQAMMQLYVLKLLKVQTKYLGRQWRKSNMKTMSAIYQKVQHRLNDDWAYGNDLDARPWDFQAEECALRANIERFNARRYDRAHSNPDFLPVDNCLQSVLGQRVDLPEDFQMNYDLWLEREVFSKPISWEELLQ | |
RUO | |
FAM40A | |
GST |
For Research Use Only