missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human Fam40A Full-Length Orf (Bab55265.1, 1 A.A.-742 A.A.) Recombinant Protein With Gst Tag At N-Terminal.

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number BAB55265.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 85369
Name FAM40A (Human) Recombinant Protein (P01)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16186303
View Documents Promotion Details
Abnova™
H00085369-P01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16196303
View Documents Promotion Details
Abnova™
H00085369-P01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Good Folding
  • Excellent amino acids labeling

Immunogen Sequence: MNRKCFEEDFRIHVTDKKWTELDTNQHRTHAMRLLDGLEVTAREKRLKVARAILYVAQGTFGECSSEAEVQSWMRYNIFLLLEVGTFNALVELLNMEIDNSAACSSAVRKPAISLADSTDLRVLLNIMYLIVETVHQECEGDKAEW

Specifications

Specifications

BAB55265.1
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer.
FAM40A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14743/KIAA1761/MGC148091/RP4-773N10.1
FAM40A
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
85369
Glutathione Sepharose 4 Fast Flow
MNRKCFEEDFRIHVTDKKWTELDTNQHRTHAMRLLDGLEVTAREKRLKVARAILYVAQGTFGECSSEAEVQSWMRYNIFLLLEVGTFNALVELLNMEIDNSAACSSAVRKPAISLADSTDLRVLLNIMYLIVETVHQECEGDKAEWRTMRQTFRAELGSPLYNNEPFAIMLFGMVTKFCSGHAPHFPMKKVLLLLWKTVLCTLGGFEELQSMKAEKRSILGLPPLPEDSIKVIRNMRAASPPASASDLIEQQQKRGRREHKALIKQDNLDAFNERDPYKADDSREEEEENDDDNSLEGETFPLERDEVMPPPLQHPQTDRLTCPKGLPWAPKVREKDIEMFLESSRSKFIGYTLGSDTNTVVGLPRPIHESIKTLKQHKYTSIAEVQAQMEEEYLRSPLSGGEEEVEQVPAETLYQGLLPSLPQYMIALLKILLAAAPTSKAKTDSINILADVLPEEMPTTVLQSMKLGVDVNRHKEVIVKAISAVLLLLLKHFKLNHVYQFEYMAQHLVFANCIPLILKFFNQNIMSYITAKNSISVLDYPHCVVHELPELTAESLEAGDSNQFCWRNLFSCINLLRILNKLTKWKHSRTMMLVVFKSAPILKRALKVKQAMMQLYVLKLLKVQTKYLGRQWRKSNMKTMSAIYQKVQHRLNDDWAYGNDLDARPWDFQAEECALRANIERFNARRYDRAHSNPDFLPVDNCLQSVLGQRVDLPEDFQMNYDLWLEREVFSKPISWEELLQ
RUO
FAM40A
GST
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only