Learn More
Abnova™ Human FADS3 Full-length ORF (NP_068373.1, 1 a.a. - 445 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_068373.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3995 |
Molecular Weight (g/mol) | 77.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137421
|
Abnova™
H00003995-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16147421
|
Abnova™
H00003995-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the fatty acid desaturase (FADS) gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13.1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization. [provided by RefSeq]
Sequence: MGGVGEPGPREGPAQPGAPLPTFCWEQIRAHDQPGDKWLVIERRVYDISRWAQRHPGGSRLIGHHGAEDATDAFRAFHQDLNFVRKFLQPLLIGELAPEEPSQDGPLNAQLVEDFRALHQAAEDMKLFDASPTFFAFLLGHILAMEVLAWLLIYLLGPGWVPSALAAFILAISQAQSWCLQHDLGHASIFKKSWWNHVAQKFVMGQLKGFSAHWWNFRHFQHHAKPNIFHKDPDVTVAPVFLLGESSVEYGKKKRRYLPYNQQHLYFFLIGPPLLTLVNFEVENLAYMLVCMQWADLLWAASFYARFFLSYLPFYGVPGVLLFFVAVRVLESHWFVWITQMNHIPKEIGHEKHRDWVSSQLAATCNVEPSLFTNWFSGHLNFQIEHHLFPRMPRHNYSRVAPLVKSLCAKHGLSYEVKPFLTALVDIVRSLKKSGDIWLDAYLHQSpecifications
NP_068373.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGGVGEPGPREGPAQPGAPLPTFCWEQIRAHDQPGDKWLVIERRVYDISRWAQRHPGGSRLIGHHGAEDATDAFRAFHQDLNFVRKFLQPLLIGELAPEEPSQDGPLNAQLVEDFRALHQAAEDMKLFDASPTFFAFLLGHILAMEVLAWLLIYLLGPGWVPSALAAFILAISQAQSWCLQHDLGHASIFKKSWWNHVAQKFVMGQLKGFSAHWWNFRHFQHHAKPNIFHKDPDVTVAPVFLLGESSVEYGKKKRRYLPYNQQHLYFFLIGPPLLTLVNFEVENLAYMLVCMQWADLLWAASFYARFFLSYLPFYGVPGVLLFFVAVRVLESHWFVWITQMNHIPKEIGHEKHRDWVSSQLAATCNVEPSLFTNWFSGHLNFQIEHHLFPRMPRHNYSRVAPLVKSLCAKHGLSYEVKPFLTALVDIVRSLKKSGDIWLDAYLHQ | |
RUO | |
FADS3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
3995 | |
FADS3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CYB5RP/LLCDL3 | |
FADS3 | |
Recombinant | |
wheat germ expression system |