Learn More
Abnova™ Human FADS3 Full-length ORF (NP_068373.1, 1 a.a. - 445 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003995-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the fatty acid desaturase (FADS) gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13.1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization. [provided by RefSeq]
Sequence: MGGVGEPGPREGPAQPGAPLPTFCWEQIRAHDQPGDKWLVIERRVYDISRWAQRHPGGSRLIGHHGAEDATDAFRAFHQDLNFVRKFLQPLLIGELAPEEPSQDGPLNAQLVEDFRALHQAAEDMKLFDASPTFFAFLLGHILAMEVLAWLLIYLLGPGWVPSALAAFILAISQAQSWCLQHDLGHASIFKKSWWNHVAQKFVMGQLKGFSAHWWNFRHFQHHAKPNIFHKDPDVTVAPVFLLGESSVEYGKKKRRYLPYNQQHLYFFLIGPPLLTLVNFEVENLAYMLVCMQWADLLWAASFYARFFLSYLPFYGVPGVLLFFVAVRVLESHWFVWITQMNHIPKEIGHEKHRDWVSSQLAATCNVEPSLFTNWFSGHLNFQIEHHLFPRMPRHNYSRVAPLVKSLCAKHGLSYEVKPFLTALVDIVRSLKKSGDIWLDAYLHQSpecifications
NP_068373.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CYB5RP/LLCDL3 | |
FADS3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3995 | |
FADS3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGGVGEPGPREGPAQPGAPLPTFCWEQIRAHDQPGDKWLVIERRVYDISRWAQRHPGGSRLIGHHGAEDATDAFRAFHQDLNFVRKFLQPLLIGELAPEEPSQDGPLNAQLVEDFRALHQAAEDMKLFDASPTFFAFLLGHILAMEVLAWLLIYLLGPGWVPSALAAFILAISQAQSWCLQHDLGHASIFKKSWWNHVAQKFVMGQLKGFSAHWWNFRHFQHHAKPNIFHKDPDVTVAPVFLLGESSVEYGKKKRRYLPYNQQHLYFFLIGPPLLTLVNFEVENLAYMLVCMQWADLLWAASFYARFFLSYLPFYGVPGVLLFFVAVRVLESHWFVWITQMNHIPKEIGHEKHRDWVSSQLAATCNVEPSLFTNWFSGHLNFQIEHHLFPRMPRHNYSRVAPLVKSLCAKHGLSYEVKPFLTALVDIVRSLKKSGDIWLDAYLHQ | |
RUO | |
FADS3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |